Isoluminol (4-Aminophthalhydrazid), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

A8264-10G A8264-1G A8264-5G
SIALA8264-10GEA 776 EUR
SIALA8264-10G SIALA8264-1G SIALA8264-5G
Isoluminol (4-Aminophthalhydrazid), Sigma-Aldrich®
Isoluminol (4-Aminophthalhydrazid)
Formel: C₈H₇N₃O₂
MDL Number: MFCD00010560
CAS-Nummer: 3682-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR