Pyridoxal-5'-phosphat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

P9255-1G P9255-5G P9255-25G
SIALP9255-1GEA 52.5 EUR
SIALP9255-1G SIALP9255-5G SIALP9255-25G
Pyridoxal-5'-phosphat, Sigma-Aldrich®
Pyridoxal-5'-phosphat
Formel: C₈H₁₀NO₆P
MDL Number: MFCD00006333
CAS-Nummer: 54-47-7
EINECS: 200-208-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR