Iminodibenzyl 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
10,11-Dihydro-5H-dibenzo[b,f]azepine
122070250
122071000
ACRO122070250EA
37.1
EUR
ACRO122070250
ACRO122071000
Iminodibenzyl 97%
Iminodibenzyl
Formel:
C₁₄H₁₃N Schmelzpunkt: 105…108 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00005070 CAS-Nummer: 494-19-9 EINECS: 207-787-1 |
Spezifikation Testergebnisse
Appearance | Yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 105 to 108 °C |
Loss on drying | ≤0.5 % |
Sulfated ash | ≤0.3 % |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...