Riboflavin-5'-phosphat Natriumsalz Dihydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Sodium (2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-3,4-dihydrobenzo[g]pteridin-10(2H)-yl)-2,3,4-trihydroxypentyl hydrogenphosphate dihydrate
F8399-10MG
F8399-1MG
F8399-5MG
SIALF8399-10MGEA
461
EUR
SIALF8399-10MG
SIALF8399-1MG
SIALF8399-5MG
Riboflavin-5'-phosphat Natriumsalz Dihydrat, Sigma-Aldrich®
Riboflavin-5'-phosphat Natriumsalz Dihydrat
Formel:
C₁₇H₂₀N₄NaO₉P·2H₂O Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00150993 CAS-Nummer: 6184-17-4 EINECS: 204-988-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...