(p-Toluolsulfonyl)methylisocyanid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-((Isocyanomethyl)sulphonyl)-4-methylbenzene, Tosylmethyl isocyanide

A14312.06 A14312.14 A14312.22
A14312.06EA 39.9 EUR
A14312.06 A14312.14 A14312.22
(p-Toluolsulfonyl)methylisocyanid 97%
(p-Toluolsulfonyl)methylisocyanid

Purity: 97+% CAS No.: 36635-61-7 Molecular Formula: C9H9NO2S Formula Weight: 195.24 Melting Point: 109-114° Beilstein Reference: 3592382 UN No.: 2811 MDL No.: MFCD00000005

Formel: C₉H₉NO₂S
Schmelzpunkt: 109…114 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00000005
CAS-Nummer: 36635-61-7
EINECS: 253-140-1
UN-Nummer: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR