tert-Butyl-N-hydroxycarbamat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine

A10349.03 A10349.06 A10349.14
A10349.03EA 33.3 EUR
A10349.03 A10349.06 A10349.14
tert-Butyl-N-hydroxycarbamat ≥98%
tert-Butyl-N-hydroxycarbamat

Purity: 98+% CAS No.: 36016-38-3 Molecular Formula: (CH3)3COCONHOH Formula Weight: 133.15 Storage Temperature: 2°C to 8°C Melting Point: 55-58° Beilstein Reference: 1756546 MDL No.: MFCD00002107

Formel: C₅H₁₁NO₃
Schmelzpunkt: 55…58 °C
MDL Number: MFCD00002107
CAS-Nummer: 36016-38-3
EINECS: 252-836-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR