Dipraseodymtricarbonat Octahydrat ≥99,9%
Lieferant: Thermo Fisher Scientific
Synonyme:
Praseodymium(III) carbonate octahydrate
14609.14
14609.22
14609.14EA
102
EUR
14609.14
14609.22
Dipraseodymtricarbonat Octahydrat ≥99,9%
Dipraseodymtricarbonat Octahydrat
Purity: 99.9%
CAS No.: 14948-62-0
Molecular Formula: Pr2(CO3)3.8H2O
Formula Weight: 605.97 (461.85anhy)
Physical Form: Crystalline
Odour: Odourless
MDL No.: MFCD00211307
Formel:
C₃O₉Pr₂·8H₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00211307 CAS-Nummer: 14948-62-0 EINECS: 227-578-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...