Acetylchlorid-[13C2] (99% ¹³C ), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ quality level = MQ200
293164-1G
293164-250MG
SIAL293164-1GEA
1050
EUR
SIAL293164-1G
SIAL293164-250MG
Acetylchlorid-[13C2] (99% ¹³C ), Sigma-Aldrich®
Acetylchlorid-[13C2]
Formel:
C₂H₃ClO |
CAS-Nummer:
89186-79-8 UN-Nummer: 1717 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...