Acetylchlorid-[1-13C] (99% ¹³C ), Sigma-Aldrich®

Lieferant: Merck
Gefahr


M-Clarity™ quality level = MQ200

293156-1G 293156-250MG
SIAL293156-1GEA 305 EUR
SIAL293156-1G SIAL293156-250MG
Acetylchlorid-[1-13C] (99% ¹³C ), Sigma-Aldrich®
Acetylchlorid-[1-13C]
Formel: C₂H₃ClO
CAS-Nummer: 1520-57-6
UN-Nummer: 1717
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR