(4-Cyanbenzyl)triphenylphosphoniumchlorid ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L12187.06
L12187.14
L12187.06EA
41.5
EUR
L12187.06
L12187.14
(4-Cyanbenzyl)triphenylphosphoniumchlorid ≥99%
(4-Cyanbenzyl)triphenylphosphoniumchlorid
Purity: 99%
CAS No.: 20430-33-5
Molecular Formula: C26H21ClNP
Formula Weight: 413.89
Odour: Odourless
Melting Point: 322-326°
Beilstein Reference: 4630137
MDL No.: MFCD00011752
Formel:
C₂₆H₂₁ClNP Schmelzpunkt: 322…326 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011752 CAS-Nummer: 20430-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...