1-Methylimidazol-4-carbonsäure

Lieferant: Apollo Scientific

OR10542-1G OR10542-5G OR10542-25G
APOSOR10542-1GEA 0 EUR
APOSOR10542-1G APOSOR10542-5G APOSOR10542-25G
1-Methylimidazol-4-carbonsäure
1-Methylimidazol-4-carbonsäure
Formel: C₅H₆N₂O₂
MDL Number: MFCD02179560
CAS-Nummer: 41716-18-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR