Imidazol-2-carbonsäureethylester ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl imidazole-2-carboxylate, Ethyl 1H-imidazole-2-carboxylate
L19405.06
L19405.03
L19405.06EA
340
EUR
L19405.06
L19405.03
Imidazol-2-carbonsäureethylester ≥97%
Imidazol-2-carbonsäureethylester
Purity: 97%
CAS No.: 33543-78-1
Molecular Formula: C6H8N2O2
Formula Weight: 140.14
Melting Point: 176-178°
Boiling Point: 48-51°/0.3mm
MDL No.: MFCD03426031
Formel:
C₆H₈N₂O₂ Molekulargewicht: 140.14 g/mol Siedepunkt: 48…51 °C (0.3 mmHg) Schmelzpunkt: 176…178 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03426031 CAS-Nummer: 33543-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...