L-Methionin-[Methyl-D3] (98% D), Sigma-Aldrich®

Lieferant: Merck

Synonyme: L-Methionine-[methyl-²H₃]

M-Clarity™ quality level = MQ200

300616-1G 300616-5G
SIAL300616-1GEA 383 EUR
SIAL300616-1G SIAL300616-5G
L-Methionin-[Methyl-D3] (98% D), Sigma-Aldrich®
L-Methionin-[Methyl-D3]
Formel: C₅H₈D₃NO₂S
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 13010-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR