5-Brom-2'-deoxyuridin (Broxuridin), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU

M-Clarity™ quality level = MQ200

B5002-100G B5002-100MG B5002-1G B5002-250MG B5002-500MG B5002-5G
SIALB5002-100GEA 9820 EUR
SIALB5002-100G SIALB5002-100MG SIALB5002-1G SIALB5002-250MG SIALB5002-500MG SIALB5002-5G
5-Brom-2'-deoxyuridin (Broxuridin), Sigma-Aldrich®
5-Brom-2'-deoxyuridin (Broxuridin)
Formel: C₉H₁₁BrN₂O₅
MDL Number: MFCD00006529
CAS-Nummer: 59-14-3
EINECS: 200-415-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR