5-Brom-2'-deoxyuridin (Broxuridin) 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU
228590010
228590025
228595000
228590100
ACRO228590010EA
89.2
EUR
ACRO228590010
ACRO228590025
ACRO228595000
ACRO228590100
5-Brom-2'-deoxyuridin (Broxuridin) 99+%
5-Brom-2'-deoxyuridin (Broxuridin)
Formel:
C₉H₁₁BrN₂O₅ Schmelzpunkt: 191…194 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00006529 CAS-Nummer: 59-14-3 EINECS: 200-415-9 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Water | ≤2 % (K.F.) |
HPLC | ≥99.0 % |
Ratio of Absorbances | 1.45 to 1.55 (A290/A260) |
Microbial contamination | ≤100 CFU/g (Total count) |
Bacterial endotoxins | ≤0.1 EU/mg (LAL-test) |
Related substances | ≤1.0 % (HPLC) Total impurities |
Specific optical rotation | +20° to +24° (20°C, 589 nm) (c=1, H2O) |
Heavy metals (as Pb) | ≤10 ppm |
Residual solvents | ≤0.1 % (GC) Methanol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...