Trihexyl(tetradecyl)phosphoniumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Trihexyl(tetradecyl)phosphonium bis(trifluoromethanesulphonyl)amide

50971-50G-F 50971-5G-F
SIAL50971-50G-FEA 261 EUR
SIAL50971-50G-F SIAL50971-5G-F
Trihexyl(tetradecyl)phosphoniumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
Trihexyl(tetradecyl)phosphoniumbis(trifluormethylsulfonyl)imid
Formel: C₃₄H₆₈F₆NO₄PS₂
Molekulargewicht: 764.00 g/mol
MDL Number: MFCD04117695
CAS-Nummer: 460092-03-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR