1,2,3,4,5-Pentachlor-6-iodbenzol

Lieferant: Thermo Scientific

BTB09644.1 BTB09644.10 BTB09644.50
MAYBBTB09644.1EA 0 EUR
MAYBBTB09644.1 MAYBBTB09644.10 MAYBBTB09644.50
1,2,3,4,5-Pentachlor-6-iodbenzol
1,2,3,4,5-Pentachlor-6-iodbenzol
Formel: C₆Cl₅I
CAS-Nummer: 16478-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR