(4-Chlorphenoxy)acetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(4-Chlorophenoxy)acetonitrile

M-Clarity™ quality level = MQ100

456861-25G 456861-5G
SIAL456861-25GEA 98.1 EUR
SIAL456861-25G SIAL456861-5G
(4-Chlorphenoxy)acetonitril, Sigma-Aldrich®
(4-Chlorphenoxy)acetonitril
Formel: C₈H₆ClNO
MDL Number: MFCD00031535
CAS-Nummer: 3598-13-8
UN-Nummer: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR