Thuliumtrinitrat Pentahydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Thulium trinitrate pentahydrate

M-Clarity™ quality level = MQ100

325996-1G 325996-5G
SIAL325996-1GEA 74.8 EUR
SIAL325996-1G SIAL325996-5G
Thuliumtrinitrat Pentahydrat, Sigma-Aldrich®
Thuliumtrinitrat Pentahydrat
Formel: N₃O₉Tm·5H₂O
MDL Number: MFCD00149875
CAS-Nummer: 36548-87-5
EINECS: 239-069-9
UN-Nummer: 1477
ADR: 5.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR