Isopropyltriphenylphosphoniumbromid

Lieferant: Thermo Scientific

RJC04032.250 RJC04032.1
MAYBRJC04032.250EA 0 EUR
MAYBRJC04032.250 MAYBRJC04032.1
Isopropyltriphenylphosphoniumbromid
Isopropyltriphenylphosphoniumbromid
Formel: C₂₁H₂₂BrP
MDL Number: MFCD00064807
CAS-Nummer: 1530-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR