Acetylchlorid-[D3], Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Acetyl chloride-[²H₃]

M-Clarity™ quality level = MQ200

175668-10G 175668-5G
SIAL175668-10GEA 316 EUR
SIAL175668-10G SIAL175668-5G
Acetylchlorid-[D3], Sigma-Aldrich®
Acetylchlorid-[D3]
Formel: C₂D₃ClO
CAS-Nummer: 19259-90-6
EINECS: 242-925-4
UN-Nummer: 1717
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR