2-Amino-3,5-dibrombenzaldehyd 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H56015.03
H56015.06
H56015.03EA
22.2
EUR
H56015.03
H56015.06
2-Amino-3,5-dibrombenzaldehyd 97%
2-Amino-3,5-dibromobenzaldehyde
Purity: 97%
CAS No.: 50910-55-9
Molecular Formula: C7H5Br2NO
Formula Weight: 278.93
Melting Point: 130-135°
MDL No.: MFCD00671100
Formel:
C₇H₅Br₂NO Siedepunkt: 331 °C (1013 hPa) Schmelzpunkt: 130…135 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00671100 CAS-Nummer: 50910-55-9 EINECS: 256-841-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...