3,5-Di-tert-butylbrenzcatechin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,5-Di-tert-butylcatechol

H55207.06 H55207.14
H55207.06EA 38.6 EUR
H55207.06 H55207.14
3,5-Di-tert-butylbrenzcatechin ≥99%
3,5-Di-tert-butylbrenzcatechin

Purity: 99% CAS No.: 1020-31-1 Molecular Formula: C14H22O2 Formula Weight: 222.33 Physical Form: Solid Appearance: White to pale brown Melting Point: 95-100° Beilstein Reference: 1370212 MDL No.: MFCD00008819

Formel: C₁₄H₂₂O₂
Siedepunkt: 310 °C (1013 hPa)
Schmelzpunkt: 95…100 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00008819
CAS-Nummer: 1020-31-1
EINECS: 213-816-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR