2'-Desoxycytidin-5'-triphosphorsäure Dinatriumsalz (dCTP-Na2), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2'-Deoxycytidine-5'-triphosphoric acid disodium salt, 2′-Deoxycytidine-5′-Triphosphate Trisodium Salt, 2'-Deoxycytidine-5'-triphosphate disodium salt
D4635-100MG
D4635-10MG
D4635-25MG
D4635-1G
SIALD4635-100MGEA
666
EUR
SIALD4635-100MG
SIALD4635-10MG
SIALD4635-25MG
SIALD4635-1G
2'-Desoxycytidin-5'-triphosphorsäure Dinatriumsalz (dCTP-Na2), Sigma-Aldrich®
2'-Desoxycytidin-5'-triphosphorsäure Dinatriumsalz (dCTP-Na2)
Formel:
C₉H₁₄N₃Na₂O₁₃P₃ |
MDL Number:
MFCD00084683 CAS-Nummer: 102783-51-7 EINECS: 252-849-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...