2'-Desoxycytidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, deoxyC

D3897-100MG D3897-250MG D3897-500MG D3897-1G D3897-5G
SIALD3897-100MGEA 29.7 EUR
SIALD3897-100MG SIALD3897-250MG SIALD3897-500MG SIALD3897-1G SIALD3897-5G
2'-Desoxycytidin, Sigma-Aldrich®
2'-Desoxycytidin
Formel: C₉H₁₃N₃O₄
MDL Number: MFCD00006547
CAS-Nummer: 951-77-9
EINECS: 213-454-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR