Inosin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
M-Clarity™ quality level = MQ200
I4125-100G
I4125-10G
I4125-1G
I4125-25G
SIALI4125-100GEA
486
EUR
SIALI4125-100G
SIALI4125-10G
SIALI4125-1G
SIALI4125-25G
Inosin, Sigma-Aldrich®
Inosin
Formel:
C₁₀H₁₂N₄O₅ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00066770 CAS-Nummer: 58-63-9 EINECS: 200-390-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...