Adenosin

Lieferant: Roth Carl

Synonyme: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside

6122.1 6122.2 6122.3
ROTH6122.1EA 0 EUR
ROTH6122.1 ROTH6122.2 ROTH6122.3
Adenosin
Adenosin
Formel: C₁₀H₁₃N₅O₄
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00005752
CAS-Nummer: 58-61-7
EINECS: 200-389-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR