Puromycin-Aminonukleosid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ300

P7130-100MG P7130-1G P7130-500MG
SIALP7130-100MGEA 775 EUR
SIALP7130-100MG SIALP7130-1G SIALP7130-500MG
Puromycin-Aminonukleosid, Sigma-Aldrich®
Puromycin-Aminonukleosid
Formel: C₁₂H₁₈N₆O₃
MDL Number: MFCD00063462
CAS-Nummer: 58-60-6
EINECS: 200-388-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR