Puromycin Dihydrochlorid
Lieferant: Roth Carl
|
Achtung
|
0240.1
0240.2
0240.3
0240.4
ROTH0240.1EA
28.8
EUR
ROTH0240.1
ROTH0240.2
ROTH0240.3
ROTH0240.4
Puromycin Dihydrochlorid
Puromycin Dihydrochlorid
Formel:
C₂₂H₂₉N₇O₅·2HCl Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00012691 CAS-Nummer: 58-58-2 EINECS: 200-387-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...