Puromycin Dihydrochlorid

Lieferant: PanReac AppliChem
Achtung

A2856.0010 A2856.0100
A2856.0010EA 53.8 EUR
A2856.0010 A2856.0100
Puromycin Dihydrochlorid
Puromycin Dihydrochlorid
Formel: C₂₂H₂₉N₇O₅·2HCl
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00012691
CAS-Nummer: 58-58-2
EINECS: 200-387-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR