Guanosin-5'-monophosphorsäure Dinatriumsalz 97%

Lieferant: Thermo Fisher Scientific

226600050 226600250
ACRO226600050EA 141 EUR
ACRO226600050 ACRO226600250
Guanosin-5'-monophosphorsäure Dinatriumsalz 97%
Guanosin-5'-monophosphorsäure Dinatriumsalz
Formel: C₁₀H₁₂N₅Na₂O₈P
Schmelzpunkt: >300 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00151238
CAS-Nummer: 5550-12-9
EINECS: 226-914-1

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤25 % (120°C, 4 h)
TLC Homogeneous
UV spectrophotometry molar absorptivity 13700 +/- 4%, pH 7
Titration with HClO4 ≥96.0 % (On dry substance)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR