2'-Deoxyguanosine monohydrate, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one hydrate

M-Clarity™ quality level = MQ200

D7145-100MG D7145-1G D7145-25MG D7145-5G
SIALD7145-100MGEA 98.6 EUR
SIALD7145-100MG SIALD7145-1G SIALD7145-25MG SIALD7145-5G
2'-Deoxyguanosine monohydrate, Sigma-Aldrich®
2'-Deoxyguanosine monohydrate
Formel: C₁₀H₁₃N₅O₄·H₂O
MDL Number: MFCD00150760
CAS-Nummer: 312693-72-4
EINECS: 213-505-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR