Ponceau S ≥75% (Farbstoffgehalt), dunkles Pulver

Lieferant: MP Biomedicals
Achtung

Synonyme: 3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt

0215194280 0215194210 0215194250
ICNA0215194280EA 206 EUR
ICNA0215194280 ICNA0215194210 ICNA0215194250
Ponceau S ≥75% (Farbstoffgehalt), dunkles Pulver
Ponceau S

Ponceau S is used as protein stain for electrophoresis.


Dye content: ~75%


Readily soluble in water.

Formel: C₂₂H₁₂N₄Na₄O₁₃S₄
Schmelzpunkt: 300 °C (lit.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003892
CAS-Nummer: 6226-79-5
EINECS: 228-319-2

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Dye Content ~75%
UV/VIS: l max (water) 517 - 523 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR