3-(4-Morpholinyl)phenol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Morpholinophenol

A15707.03 A15707.06 A15707.14
A15707.03EA 40.8 EUR
A15707.03 A15707.06 A15707.14
3-(4-Morpholinyl)phenol ≥98%
3-(4-Morpholinyl)phenol

Purity: 98% CAS No.: 27292-49-5 Molecular Formula: C10H13NO2 Formula Weight: 179.22 Melting Point: 127-132° Beilstein Reference: 149989

Formel: C₁₀H₁₃NO₂
Schmelzpunkt: 127…132 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00051675
CAS-Nummer: 27292-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR