MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure) ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-3-morpholinopropane-1-sulphonic acid
44135.18
44135.30
44135.A1
44135.18EA
79.8
EUR
44135.18
44135.30
44135.A1
MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure) ≥98%
MOPSO (3-(4-Morpholino)-2-hydroxypropansulfonsäure)
Purity: 98%
CAS No.: 68399-77-9
Molecular Formula: C7H15NO5S
Formula Weight: 225.27
Physical Form: Powder
Melting Point: 276-279°
MDL No.: MFCD00009629
Biological buffers, pH range 6.2-7.6
Formel:
C₇H₁₅NO₅S Schmelzpunkt: 276…279 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00009629 CAS-Nummer: 68399-77-9 EINECS: 269-989-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...