Kaliumxanthogenat (Kalium-O-ethyldithiocarbonat) ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Potassium-O-ethyldithiocarbonate, Potassium ethyl xanthate, Ethylxanthic acid potassium salt
Purity: 98%
CAS No.: 140-89-6
Molecular Formula: C3H5KOS2
Formula Weight: 160.30
Flash Point: 205°(401°F)
Physical Form: Powder
Density: 1.558
Appearance: Yellow
Odour: Unpleasant
pH: 9-10
Melting Point: ca 205° dec.
Beilstein Reference: 3596974
Merck Reference: 14,7696
MDL No.: MFCD00004931
Formel:
C₃H₅KOS₂ Schmelzpunkt: ∼ 205 °C (dec.) Dichte: 1.558 Flammpunkt: 205 °C (401 °F) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004931 CAS-Nummer: 140-89-6 EINECS: 205-439-3 Merck Index: 14,7696 |