Rhodamin 6G Perchlorat 96%

Lieferant: Thermo Fisher Scientific
Achtung

H33626.MD H33626.03
H33626.MDEA 38.9 EUR
H33626.MD H33626.03
Rhodamin 6G Perchlorat 96%
Rhodamin 6G Perchlorat

Purity: 96% CAS No.: 13161-28-9 Molecular Formula: C28H31ClN2O7 Formula Weight: 543.02 Melting Point: 240° MDL No.: MFCD00013117

Formel: C₂₈H₃₁ClN₂O₇
Molekulargewicht: 543.02 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013117
CAS-Nummer: 13161-28-9
EINECS: 236-099-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR