1-Ethyl-3-methylimidazolium-p-toluolsulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethyl-3-methylimidazolium tosylate, 1-Ethyl-3-methyl-1H-imidazol-3-ium 4-methylbenzenesulphonate, [EMIM] [TOS]

89155-50G-F 89155-5G-F
SIAL89155-50G-FEA 572 EUR
SIAL89155-50G-F SIAL89155-5G-F
1-Ethyl-3-methylimidazolium-p-toluolsulfonat, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium-p-toluolsulfonat
Formel: C₁₃H₁₈N₂O₃S
MDL Number: MFCD03788929
CAS-Nummer: 328090-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR