Azur B (Azur I)

Lieferant: Merck

Synonyme: C.I. 52010, N,N,N′-Trimethylthionin, Azure B, Trimethylthionine chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chloride, Methylene Azur B

2.79200.0000 2.79200.0500 2.79200.2500
2.79200.0000EA 0 EUR
2.79200.0000 2.79200.0500 2.79200.2500
Azur B (Azur I)
Azur B (Azur I)
Formel: C₁₅H₁₆ClN₃S
Molekulargewicht: 305.83 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011935
CAS-Nummer: 531-55-5
EINECS: 208-511-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR