Alizarinrot S (Natriumsalz)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Sodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate, C.I. 58005, Alizarin sulphonic acid disodium salt, Alizarin Red S sodium

42040.06 42040.14
42040.06EA 43.5 EUR
42040.06 42040.14
Alizarinrot S (Natriumsalz)
Alizarinrot S (Natriumsalz)

CAS No.: 130-22-3 Molecular Formula: C14H7O7SNa Formula Weight: 342.27 Physical Form: Powder Appearance: Yellow to red Beilstein Reference: 4117090 Merck Reference: 14,8573 MDL No.: MFCD00013049

Formel: C₁₄H₇NaO₇S
Molekulargewicht: 342.26 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013049
CAS-Nummer: 130-22-3
EINECS: 204-981-8
Merck Index: 14,8573

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR