3,5-Di-tert-butyl-2-hydroxybenzaldehyd ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,5-Di-tert-butylsalicylaldehyde
B23834.06
B23834.14
B23834.22
B23834.06EA
55.8
EUR
B23834.06
B23834.14
B23834.22
3,5-Di-tert-butyl-2-hydroxybenzaldehyd ≥99%
3,5-Di-tert-butyl-2-hydroxybenzaldehyd
Purity: 99%
CAS No.: 37942-07-7
Molecular Formula: C15H22O2
Formula Weight: 234.34
Physical Form: Cryst solid
Sensitivity: Air Sensitive
Beilstein Reference: 1877810
MDL No.: MFCD00191998
Formel:
C₁₅H₂₂O₂ Molekulargewicht: 234.34 g/mol Schmelzpunkt: 59…61 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00191998 CAS-Nummer: 37942-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...