3,5-Dibrom-2-fluorpyridin

Lieferant: Apollo Scientific

PC8845-25G PC8845-100G
APOSPC8845-25GEA 58 EUR
APOSPC8845-25G APOSPC8845-100G
3,5-Dibrom-2-fluorpyridin
3,5-Dibromo-2-fluoropyridine
Formel: C₅H₂Br₂FN
MDL Number: MFCD07368619
CAS-Nummer: 473596-07-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR