4-Brom-6-chlor-o-kresol

Lieferant: Thermo Scientific

Synonyme: 4-Bromo-2-chloro-6-methylphenol

SB01216DA SB01216EA SB01216EE
MAYBSB01216DAEA 0 EUR
MAYBSB01216DA MAYBSB01216EA MAYBSB01216EE
4-Brom-6-chlor-o-kresol
4-Brom-6-chlor-o-kresol
Formel: C₇H₆BrClO
MDL Number: MFCD00002167
CAS-Nummer: 7530-27-0
EINECS: 231-394-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR