Guanosin ≥98%
Lieferant: Thermo Fisher Scientific
A11328.18
A11328.14
A11328.22
A11328.30
A11328.18EA
110
EUR
A11328.18
A11328.14
A11328.22
A11328.30
Guanosin ≥98%
Guanosin
Purity: 98+%
CAS No.: 118-00-3
Molecular Formula: C10H13N5O5
Formula Weight: 283.24
Melting Point: ca 250° dec.
Beilstein Reference: 625911
Merck Reference: 14,4566
Optical Rotation: -72° (c=1.5 in 0.1N NaOH)
MDL No.: MFCD00010182
Formel:
C₁₀H₁₃N₅O₅ Schmelzpunkt: ∼ 250 °C (Zers.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00010182 CAS-Nummer: 118-00-3 EINECS: 204-227-8 Merck Index: 14,4566 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...