Betain, wasserfrei 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
TMG, Trimethylglycine, 2-(Trimethylammonio)acetate
B24397.0E
B24397.22
B24397.36
B24397.0EEA
534
EUR
B24397.0E
B24397.22
B24397.36
Betain, wasserfrei 98%
Betain
Purity: 98%
CAS No.: 107-43-7
Molecular Formula: (CH3)3NCH2CO2
Formula Weight: 117.15
Physical Form: Cryst powder
Sensitivity: Hygroscopic
Appearance: White
Melting Point:
Formel:
C₅H₁₁NO₂ Molekulargewicht: 117.15 g/mol Schmelzpunkt: >300 °C (Zers.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012123 CAS-Nummer: 107-43-7 EINECS: 203-490-6 Merck Index: 14,1179 |