Betain Monohydrat

Lieferant: BIOMOL RESEARCH LABORATORIES

53920-1 53920-2
BMOL53920-1EA 0 EUR
BMOL53920-1 BMOL53920-2
Betain Monohydrat
Betain Monohydrat
Formel: C₅H₁₁NO₂·1H₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00150010
CAS-Nummer: 590-47-6
EINECS: 203-490-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR