Sicherheitsschuhe mit Klettverschluss, GX 350
Lieferant: ATLAS - the shoe company
45535 45536 45537 45538 45539 45540 45541 45542 45543
114-1376EA 0 EUR
114-1376 114-1377 114-1378 114-1379 114-1380 114-1381 114-1382 114-1383 114-1384
Sicherheitsschuhe mit Klettverschluss, GX 350
Schuhe und Stiefel
Safety shoes made from Sportline upper (oiled nubuck leather), reflectors and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
  • Electrical contact resistance of 1×10⁵ to 3,5×10⁷ Ohm
Farbe: Dunkelblau/Grau
Colour: Black/blue
Zertifizierungen: DGUV 112-191; EN ISO 20345; S1, SRC