Ampicillin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
A9393-5G
A9393-25G
SIALA9393-5GEA
84.6
EUR
SIALA9393-5G
SIALA9393-25G
Ampicillin, Sigma-Aldrich®
Ampicillin
Formel:
C₁₆H₁₉N₃O₄S Molekulargewicht: 349.4 g/mol |
MDL Number:
MFCD00005175 CAS-Nummer: 69-53-4 EINECS: 200-709-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...