Electrophoresis gel plate racks, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F135950000 F135960000
581-7611EA 282 EUR
581-7611 BELAF135960000
Electrophoresis gel plate racks, SP Bel-Art
Gestelle Gestelle für Gelelektrophorese-Platten
Versatile solution for drying and storing gel plates. Rugged, A-shaped racks that securely supports glass gel plates in an upright position for rapid drying and easy access.
  • 10 plate capacity