γ-Cyclodextrin Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: γ-Cyclodextrin hydrate, γ-Cyclodextrin xhydrate

M-Clarity™ quality level = MQ100

861413-100MG 861413-1G
SIAL861413-100MGEA 53.1 EUR
SIAL861413-100MG SIAL861413-1G
γ-Cyclodextrin Hydrat, Sigma-Aldrich®
γ-Cyclodextrin Hydrat
Formel: C₄₈H₈₀O₄₀·H₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00149574
CAS-Nummer: 91464-90-3
EINECS: 241-482-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR